3-Methylquinoline ≥98.0% (by GC)

Supplier: TCI America
Warning

M0782-25G M0782-500G
TCM0782-025GEA 213.3 CAD
TCM0782-025G TCM0782-500G
3-Methylquinoline ≥98.0% (by GC)
3-Methylquinoline

CAS Number: 612-58-8
MDL Number: MFCD00014661
Molecular Formula: C10H9N
Molecular Weight: 143.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 255
Flash Point (°C): 113
Freezing point (°C): 15
Specific Gravity (20/20): 1.07
Lambda max.: 318 nm (EtOH aq.)

Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 255 °C
Melting Pt: 17 °C
Density: 1.07 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00014661
CAS Number: 612-58-8
EINECS: 210-315-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=199c6f06f009c3cf8b3ebecd1c43afb4b1083438&vid=e22a6e9a4a594e0e25d8a8a297574b942b1d99be&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White or Colourless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR