6-Ethyl-o-toluidine ≥98.0%
Supplier: TCI America
Synonyms:
2-Ethyl-6-methylaniline, 2-Amino-3-ethyltoluene
M0772-25ML
M0772-500ML
TCM0772-025MLEA
44.24
CAD
TCM0772-025ML
TCM0772-500ML
6-Ethyl-o-toluidine ≥98.0%
6-Ethyl-o-toluidine
CAS Number: 24549-06-2
MDL Number: MFCD00007752
Molecular Formula: C9H13N
Molecular Weight: 135.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 231
Melting point (°C): -33
Flash Point (°C): 111
Specific Gravity (20/20): 0.97
Other Product Names / Synonyms: 2-Ethyl-6-methylaniline, 6-Ethyl-o-toluidine
Formula:
C₉H₁₃N MW: 135.21 Boiling Pt: 231 Melting Pt: -33 Density: 0.97 (20 °C) Flash Pt: 111 °C |
MDL Number:
MFCD00007752 CAS Number: 24549-06-2 EINECS: 246-309-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...