4'-Methylpropiophenone ≥95.0%

Supplier: TCI America

Synonyms: p-Methylpropiophenone

M0768-25ML M0768-500ML
TCM0768-025MLEA 56.25 CAD
TCM0768-025ML TCM0768-500ML
4'-Methylpropiophenone ≥95.0%
4'-Methylpropiophenone

CAS Number: 5337-93-9
MDL Number: MFCD00009312
Molecular Formula: C10H12O
Molecular Weight: 148.21
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 239
Flash Point (°C): 109
Specific Gravity (20/20): 0.99

Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 239 °C
Melting Pt: 91 °C
Density: 0.99 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD00009312
CAS Number: 5337-93-9
EINECS: 226-267-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2bdcef2d87617511635beaf113c55e7d424dafc&vid=c2bdcef2d87617511635beaf113c55e7d424dafc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR