7-Amino-4-methylcoumarin ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M0760-1G M0760-5G
TCM0760-001GEA 214.88 CAD
TCM0760-001G TCM0760-5G
7-Amino-4-methylcoumarin ≥98.0% (by HPLC, titration analysis)
7-Amino-4-methylcoumarin

[for HPLC Labeling]
CAS Number: 26093-31-2
MDL Number: MFCD00006868
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 228

Formula: C₁₀H₉NO₂
Boiling Pt: 352 °C (1013 hPa)
Melting Pt: 228 °C
Density: 1.377 g/cm³ (20 °C)
MDL Number: MFCD00006868
CAS Number: 26093-31-2
EINECS: 247-454-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c17d12907b6d98bfa0a9aa60e2fba61d0d1fa99&vid=d7542253a3a9274379f527a50147d0402a4202a7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR