MOPS-Na ≥98.0%

Supplier: TCI America

Synonyms: 3-(N-Morpholino)propanesulphonic acid sodium salt, Sodium 3-morpholinopropane-1-sulphonate, Sodium 4-morpholin-1-ylpropylsulphonate, 3-(N-Morpholino)propanesulfonic acid sodium salt, Sodium 3-morpholinopropane-1-sulfonate, MOPS sodium salt, Sodium 4-morpholin-1-ylpropylsulfonate

M0755-250G M0755-25G
TCM0755-250GEA 295.93 CAD
TCM0755-250G TCM0755-025G
MOPS-Na ≥98.0%
MOPS-Na

CAS Number: 71119-22-7
MDL Number: MFCD00064350
Molecular Formula: C7H15NO4S
Molecular Weight: 231.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: MOPS Sodium Salt, 3-Morpholinopropanesulfonic Acid Sodium Salt

Formula: C₇H₁₄NNaO₄S
MW: 231.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064350
CAS Number: 71119-22-7
EINECS: 428-420-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1e60c1fa2f5cbfa2202e7039231ce310eade30e&vid=b1e60c1fa2f5cbfa2202e7039231ce310eade30e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR