3,4-(Methylenedioxy)aniline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M0741-25G
M0741-100G
TCM0741-025GEA
135.88
CAD
TCM0741-025G
TCM0741-100G
3,4-(Methylenedioxy)aniline ≥98.0% (by GC, titration analysis)
3,4-(Methylenedioxy)aniline
CAS Number: 14268-66-7
MDL Number: MFCD00005832
Molecular Formula: C7H7NO2
Molecular Weight: 137.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 156
Melting point (°C): 44
Flash Point (°C): 142
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 5-Amino-1,3-benzodioxole, 4-Amino-1,2-methylenedioxybenzene
Formula:
C₇H₇NO₂ Boiling Pt: 156 °C Melting Pt: 44 °C Flash Pt: 142 °C |
MDL Number:
MFCD00005832 CAS Number: 14268-66-7 EINECS: 238-161-6 UN: 2811 ADR: 6.1,III |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...