4-(Methylthio)benzaldehyde ≥97.0%
Supplier: TCI America
M0739-25G
M0739-250G
TCM0739-025GEA
69.52
CAD
TCM0739-025G
TCM0739-250G
4-(Methylthio)benzaldehyde ≥97.0%
4-(Methylthio)benzaldehyde
CAS Number: 3446-89-7
MDL Number: MFCD00006948
Molecular Formula: C8H8OS
Molecular Weight: 152.21
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 153
Flash Point (°C): 110
Specific Gravity (20/20): 1.17
Other Product Names / Synonyms: 4-Methylmercaptobenzaldehyde
Formula:
C₈H₈OS Boiling Pt: 153 °C Density: 1.17 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00006948 CAS Number: 3446-89-7 EINECS: 222-365-7 UN: 3334 ADR: 9, |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...