1-Methyl-2-phenylindole ≥99.0% (by total nitrogen basis)

Supplier: TCI America

M0733-25G M0733-500G
TCM0733-025GEA 79 CAD
TCM0733-025G TCM0733-500G
1-Methyl-2-phenylindole ≥99.0% (by total nitrogen basis)
1-Methyl-2-phenylindole

CAS Number: 3558-24-5
MDL Number: MFCD00022892
Molecular Formula: C15H13N
Molecular Weight: 207.28
Purity/Analysis Method: >99.0% (N)
Form: Crystal
Melting point (°C): 102

Formula: C₁₅H₁₃N
Melting Pt: 102 °C
MDL Number: MFCD00022892
CAS Number: 3558-24-5
EINECS: 222-618-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eb6a4bbdec45af0a102312d9f8fe02debb19b0d1&vid=eb6a4bbdec45af0a102312d9f8fe02debb19b0d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR