1-Methylcyclohexanecarboxylic acid ≥99.0% (by GC, titration analysis)
Supplier: TCI America
M0732-25G
M0732-5G
TCM0732-025GEA
200.66
CAD
TCM0732-025G
76220-228
1-Methylcyclohexanecarboxylic acid ≥99.0% (by GC, titration analysis)
1-Methylcyclohexanecarboxylic acid
CAS Number: 1123-25-7
MDL Number: MFCD00001463
Molecular Formula: C8H14O2
Molecular Weight: 142.20
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Boiling point (°C): 236
Melting point (°C): 38
Flash Point (°C): 100
Formula:
C₈H₁₄O₂ Boiling Pt: 236 °C Melting Pt: 38 °C Density: 1.0115 g/cm³ (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00001463 CAS Number: 1123-25-7 EINECS: 214-371-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...