5-Methoxy-2-methylindole ≥99.0%
Supplier: TCI America
Synonyms:
5-Methoxy-2-methyl-1H-indole
M0730-1G
M0730-5G
TCM0730-001GEA
233.84
CAD
TCM0730-001G
TCM0730-5G
5-Methoxy-2-methylindole ≥99.0%
5-Methoxy-2-methylindole
CAS Number: 1076-74-0
MDL Number: MFCD00005620
Molecular Formula: C10H11NO
Molecular Weight: 161.20
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 87
Formula:
C₁₀H₁₁NO MW: 161.2 g/mol Boiling Pt: 145 Melting Pt: 89 |
MDL Number:
MFCD00005620 CAS Number: 1076-74-0 EINECS: 214-066-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...