3-(Trimethoxysilyl)propyl methacrylate ≥98.0% stabilized

Supplier: TCI America


Stabilization: Stabilised with Stabilised

M0725-25ML M0725-500ML M0725-100ML
TCM0725-025MLEA 51.41 CAD
TCM0725-025ML TCM0725-500ML TCM0725-100ML
3-(Trimethoxysilyl)propyl methacrylate ≥98.0% stabilized
3-(Trimethoxysilyl)propyl methacrylate

(stabilized with BHT)
CAS Number: 2530-85-0
MDL Number: MFCD00008593
Molecular Formula: C10H20O5Si
Molecular Weight: 248.35
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 92
Specific Gravity (20/20): 1.05


Product Categories: Materials Chemistry, Silane Coupling Agents, Monomers
Other Product Names / Synonyms: Methacrylic Acid 3-(Trimethoxysilyl)propyl Ester, 3-(Methacryloyloxy)propyltrimethoxysilane

Formula: C₁₀H₂₀O₅Si
Boiling Pt: 110 °C
Density: 1.05 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00008593
CAS Number: 2530-85-0
EINECS: 219-785-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0fa0a2ea1c3ebfb5052e07471def13563d0c246f&vid=0fa0a2ea1c3ebfb5052e07471def13563d0c246f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR