Ethyl-2-methylacetoacetate ≥95.0%
Supplier: TCI America
M0698-250ML
M0698-25ML
TCM0698-250MLEA
324.75
CAD
TCM0698-250ML
TCM0698-025ML
Ethyl-2-methylacetoacetate ≥95.0%
Ethyl 2-methylacetoacetate
CAS Number: 609-14-3
MDL Number: MFCD00009164
Molecular Formula: C7H12O3
Molecular Weight: 144.17
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 185
Flash Point (°C): 62
Specific Gravity (20/20): 1.00
Other Product Names / Synonyms: 2-Acetylpropionic Acid Ethyl Ester, Ethyl 2-Acetylpropionate, 2-Methylacetoacetic Acid Ethyl Ester
Formula:
C₇H₁₂O₃ Boiling Pt: 185 °C Melting Pt: –45 °C Density: 1.00 (20 °C) Flash Pt: 62 °C |
MDL Number:
MFCD00009164 CAS Number: 609-14-3 EINECS: 210-179-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...