3-Methoxy-3-methyl-1-butanol ≥98.0%

Supplier: TCI America

Synonyms: 3-Methyl-3-methoxybutanol, Solvant 39, MMB, 3-Methoxy-3-methylbutan-1-ol

M0695-500ML M0695-25ML
TCM0695-500MLEA 71.1 CAD
TCM0695-500ML TCM0695-025ML
3-Methoxy-3-methyl-1-butanol ≥98.0%
3-Methoxy-3-methyl-1-butanol

CAS Number: 56539-66-3
MDL Number: MFCD00044771
Molecular Formula: C6H14O2
Molecular Weight: 118.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 174
Flash Point (°C): 67
Specific Gravity (20/20): 0.93

Formula: C₆H₁₄O₂
MW: 118.18 g/mol
Boiling Pt: 174 °C
Melting Pt: –50 °C
Density: 0.93 (20 °C)
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00044771
CAS Number: 56539-66-3
EINECS: 260-252-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c52ee271362930be4b3c9fb1ef5affcfb00d165&vid=1c52ee271362930be4b3c9fb1ef5affcfb00d165&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR