3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
M0687-25G
M0687-500G
TCM0687-025GEA
86.38
CAD
TCM0687-025G
TCM0687-500G
3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98.0% (by HPLC, titration analysis)
3-Methyl-1-phenyl-2-pyrazolin-5-one
CAS Number: 89-25-8
MDL Number: MFCD00003138
Molecular Formula: C10H10N2O
Molecular Weight: 174.20
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 287
Melting point (°C): 129
Product Categories: Bioscience, Other Pharmaceutical Ingredients, Reagents for Nitric Oxide Research
Other Product Names / Synonyms: Monopyrazolone, Edaravone
Formula:
C₁₀H₁₀N₂O Boiling Pt: 287 °C Melting Pt: 129 °C |
MDL Number:
MFCD00003138 CAS Number: 89-25-8 EINECS: 201-891-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...