4-Methoxyphenylacetone ≥97.0%
Supplier: TCI America
M0675-25ML
M0675-100ML
TCM0675-025MLEA
77.23
CAD
TCM0675-025ML
TCM0675-100ML
4-Methoxyphenylacetone ≥97.0%
4-Methoxyphenylacetone
CAS Number: 122-84-9
MDL Number: MFCD00008773
Molecular Formula: C10H12O2
Molecular Weight: 164.20
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 269
Flash Point (°C): 101
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: 4-Acetonylanisole
Formula:
C₁₀H₁₂O₂ Boiling Pt: 269 °C Melting Pt: –15 °C Density: 1.07 (20 °C) Flash Pt: 101 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008773 CAS Number: 122-84-9 EINECS: 204-578-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...