1-(Hydroxymethyl)-5,5-dimethylhydantoin ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

M0668-25G M0668-500G
TCM0668-025GEA 53.72 CAD
TCM0668-025G TCM0668-500G
1-(Hydroxymethyl)-5,5-dimethylhydantoin ≥98.0% (by HPLC, titration analysis)
1-(Hydroxymethyl)-5,5-dimethylhydantoin

CAS Number: 116-25-6
MDL Number: MFCD00020866
Molecular Formula: C6H10N2O3
Molecular Weight: 158.16
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Other Product Names / Synonyms: 1-Methylol-5,5-dimethylhydantoin

Formula: C₆H₁₀N₂O₃
MDL Number: MFCD00020866
CAS Number: 116-25-6
EINECS: 204-132-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9e337463cdd7b0041f8f4b27064a93f7b3c358b8&vid=9e337463cdd7b0041f8f4b27064a93f7b3c358b8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR