(R)-(-)-Mandelic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

M0662-25G M0662-500G M0662-100G
TCM0662-025GEA 104.49 CAD
TCM0662-025G TCM0662-500G TCM0662-100G
(R)-(-)-Mandelic acid ≥99.0% (by GC, titration analysis)
(R)-(-)-Mandelic acid

CAS Number: 611-71-2
MDL Number: MFCD00064251
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 134
Specific rotation [a]20/D: -153.5 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (R)-(-)-Amygdalic Acid

Formula: C₈H₈O₃
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 134 °C
Density: 1.341 g/cm³ (20 °C)
Flash Pt: >190 °C
MDL Number: MFCD00064251
CAS Number: 611-71-2
EINECS: 210-276-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=644a09de7bbeb4da749535736d2f891f12d2e57a&vid=644a09de7bbeb4da749535736d2f891f12d2e57a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR