(S)-(+)-Mandelic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America
Danger

M0661-25G M0661-250G
TCM0661-025GEA 129.56 CAD
TCM0661-025G TCM0661-250G
(S)-(+)-Mandelic acid ≥99.0% (by GC, titration analysis)
(S)-(+)-Mandelic acid

CAS Number: 17199-29-0
MDL Number: MFCD00004495
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 134
Flash Point (°C): 200
Specific rotation [a]20/D: 153.5 deg (C=1, H2O)


Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks, Analytical Science, e.e. / Absolute Configuration Determination (NMR)
Other Product Names / Synonyms: (S)-(+)-Amygdalic Acid

Formula: C₈H₈O₃
Boiling Pt: 322 °C (1013 hPa)
Melting Pt: 134 °C
Density: 1.349 g/cm³ (20 °C)
Flash Pt: 200 °C
MDL Number: MFCD00004495
CAS Number: 17199-29-0
EINECS: 241-240-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=740f275191c4efc81631cb30e369f39124ce93b3&vid=740f275191c4efc81631cb30e369f39124ce93b3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR