7-(Diethylamino)-4-methylcoumarin ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Coumarin 1, 7-(Diethylamino)-4-methyl-2H-chromen-2-one, 7-Diethylamino-4-methylcoumarin

M0631-500G M0631-25G
TCM0631-500GEA 461.36 CAD
TCM0631-500G TCM0631-025G
7-(Diethylamino)-4-methylcoumarin ≥98.0% (by GC, titration analysis)
7-(Diethylamino)-4-methylcoumarin

CAS Number: 91-44-1
MDL Number: MFCD00006864
Molecular Formula: C14H17NO2
Molecular Weight: 231.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 240
Melting point (°C): 72
Flash Point (°C): 152

Formula: C₁₄H₁₇NO₂
MW: 231.30 g/mol
Boiling Pt: 240 °C
Melting Pt: 72 °C
Flash Pt: 152 °C
MDL Number: MFCD00006864
CAS Number: 91-44-1
EINECS: 202-068-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47b8ea54b92023d92718633f89fd3532c71cea4f&vid=47b8ea54b92023d92718633f89fd3532c71cea4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR