2-Ethylhexyl methacrylate ≥99.0% stabilized

Supplier: TCI America


Stabilization: Stabilised with Stabilised

M0591-25ML M0591-500ML
TCM0591-025MLEA 30.02 CAD
TCM0591-025ML TCM0591-500ML
2-Ethylhexyl methacrylate ≥99.0% stabilized
2-Ethylhexyl methacrylate

(stabilized with MEHQ)
CAS Number: 688-84-6
MDL Number: MFCD00009494
Molecular Formula: C12H22O2
Molecular Weight: 198.31
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 100
Specific Gravity (20/20): 0.88


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Methacrylic Acid 2-Ethylhexyl Ester, Methacrylic Acid Octyl Ester, Octyl Methacrylate

Formula: C₁₂H₂₂O₂
Boiling Pt: 88 °C
Density: 0.88 (20 °C)
Flash Pt: 100 °C
MDL Number: MFCD00009494
CAS Number: 688-84-6
EINECS: 211-708-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3fb70c6c10886ba585a9150688ba029afbd12c2&vid=a3fb70c6c10886ba585a9150688ba029afbd12c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR