α-Methylcinnamaldehyde ≥95.0%

Supplier: TCI America

M0584-25G M0584-500G
TCM0584-025GEA 170.43 CAD
TCM0584-025G TCM0584-500G
α-Methylcinnamaldehyde ≥95.0%
α-Methylcinnamaldehyde

CAS Number: 101-39-3
MDL Number: MFCD00006976
Molecular Formula: C10H10O
Molecular Weight: 146.19
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 132
Flash Point (°C): 124
Specific Gravity (20/20): 1.04


Other Product Names / Synonyms: 2-Methyl-3-phenylacrolein, 3-Phenylmethacrolein, 2-Methyl-3-phenyl-2-propenal

Formula: C₁₀H₁₀O
Boiling Pt: 132 °C
Density: 1.04 (20 °C)
Flash Pt: 124 °C
MDL Number: MFCD00006976
CAS Number: 101-39-3
EINECS: 202-938-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2008156c800f6d34248214682ae76a4dcedef32d&vid=2008156c800f6d34248214682ae76a4dcedef32d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR