2-Methylquinoxaline ≥98.0%
Supplier: TCI America
|
Warning
|
M0580-5G
M0580-25G
TCM0580-005GEA
63.2
CAD
TCM0580-005G
TCM0580-025G
2-Methylquinoxaline ≥98.0%
2-Methylquinoxaline
CAS Number: 7251-61-8
MDL Number: MFCD00006727
Molecular Formula: C9H8N2
Molecular Weight: 144.18
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 245
Flash Point (°C): 107
Specific Gravity (20/20): 1.12
Formula:
C₉H₈N₂ Boiling Pt: 245 °C Density: 1.12 (20 °C) Flash Pt: 107 °C |
MDL Number:
MFCD00006727 CAS Number: 7251-61-8 EINECS: 230-664-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...