3-Methyl-4-nitrobenzoic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3-Methyl-4-nitrobenzoic acid, 4-Nitro-m-toluic acid

M0565-500G M0565-25G
TCM0565-500GEA 592.5 CAD
TCM0565-500G TCM0565-025G
3-Methyl-4-nitrobenzoic acid ≥98.0% (by HPLC, titration analysis)
4-Nitro-m-toluic acid

CAS Number: 3113-71-1 MDL Number: MFCD00007168 Molecular Formula: C8H7NO4 Molecular Weight: 181.15 Purity/Analysis Method: 98.0% (T,HPLC) Form: Crystal Melting point (°C): 217


Form: Crystal
Melting point (°C): 217

Formula: C₈H₇NO₄
MW: 181.15 g/mol
Boiling Pt: 356
Melting Pt: 217 °C
Storage Temperature: Ambient
MDL Number: MFCD00007168
CAS Number: 3113-71-1
EINECS: 221-479-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d17d75f5d6c0cf742b806a9116829ed8eb1a23cd&vid=12dc071ede2dbcbfcde21f4f803b83bb30e17e18&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T,HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR