4-Methylbiphenyl ≥98.0%

Supplier: TCI America

Synonyms: 4-Methyl-1,1'-biphenyl, 4-Phenyltoluene

M0563-5G M0563-25G
TCM0563-005GEA 155.31 CAD
TCM0563-005G TCM0563-025G
4-Methylbiphenyl ≥98.0%
4-Methylbiphenyl

CAS Number: 644-08-6
MDL Number: MFCD00008544
Molecular Formula: C13H12
Molecular Weight: 168.24
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 268
Melting point (°C): 47
Flash Point (°C): 110


Other Product Names / Synonyms: 4-Phenyltoluene, p-Tolylbenzene

Formula: C₁₃H₁₂
Boiling Pt: 268 °C
Melting Pt: 47 °C
Density: 1.015 g/cm³ (27 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008544
CAS Number: 644-08-6
EINECS: 211-409-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6df4b599e2015f0800b0d2a79ce86362df4385b5&vid=cb367f6dddc55e49a696bb0989bc4606da75f4da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR