1-Methylindole ≥96.0%

Supplier: TCI America

Synonyms: N-Methylindole, 1-Methyl-1H-indole

M0561-5G M0561-25G
TCM0561-005GEA 31.84 CAD
TCM0561-005G TCM0561-025G
1-Methylindole ≥96.0%
1-Methylindole

CAS Number: 603-76-9
MDL Number: MFCD00005800
Molecular Formula: C9H9N
Molecular Weight: 131.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 246
Flash Point (°C): 110
Specific Gravity (20/20): 1.05

Formula: C₉H₉N
MW: 131.18 g/mol
Boiling Pt: 246 °C
Melting Pt: 94…95 °C
Density: 1.05 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00005800
CAS Number: 603-76-9
EINECS: 210-057-5
UN: 3334
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f2d218de32e35b25dc1b2826454a6cf3b78a05d4&vid=f2d218de32e35b25dc1b2826454a6cf3b78a05d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR