N-Methylformanilide ≥99.0%
Supplier: TCI America
M0552-25G
M0552-500G
M0552-100G
TCM0552-025GEA
28.28
CAD
TCM0552-025G
TCM0552-500G
TCM0552-100G
N-Methylformanilide ≥99.0%
N-Methylformanilide
CAS Number: 93-61-8
MDL Number: MFCD00003283
Molecular Formula: C8H9NO
Molecular Weight: 135.17
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 243
Flash Point (°C): 129
Freezing point (°C): 14
Specific Gravity (20/20): 1.10
Product Categories: Synthetic Organic Chemistry, Formylation
Other Product Names / Synonyms: N-Formyl-N-methylaniline
Formula:
C₈H₉NO Boiling Pt: 243 °C Density: 1.10 (20 °C) Flash Pt: 129 °C |
MDL Number:
MFCD00003283 CAS Number: 93-61-8 EINECS: 202-262-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...