Potassium methanesulfonate ≥98.0%

Supplier: TCI America

Synonyms: Potassiummethanesulfonate, Potassiummethanesulphonate, Methanesulphonic acid potassium salt, Methanesulfonic acid potassium salt

M0550-25G M0550-100G
TCM0550-025GEA 77.42 CAD
TCM0550-025G TCM0550-100G
Potassium methanesulfonate ≥98.0%
Potassium methanesulphonate

CAS Number: 2386-56-3
MDL Number: MFCD00070544
Molecular Formula: CH4O3S
Molecular Weight: 134.19
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Methanesulfonic Acid Potassium Salt

Formula: CH₃KO₃S
MW: 134.2 g/mol
MDL Number: MFCD00070544
CAS Number: 2386-56-3
EINECS: 219-202-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9ce91be3a01afea41566bf07cd032dd8b9c62634&vid=9ce91be3a01afea41566bf07cd032dd8b9c62634&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR