Diethylene glycol monomethyl ether ≥99.0% stabilised
Supplier: TCI America
|
Warning
|
Synonyms:
2-(2-Methoxyethoxy)ethanol, Diethylene glycol methyl ether
(stabilized with BHT)
CAS Number: 111-77-3
MDL Number: MFCD00002871
Molecular Formula: C5H12O3
Molecular Weight: 120.15
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 193
Melting point (°C): -70
Flash Point (°C): 105
Specific Gravity (20/20): 1.02
Product Categories: Bioscience, PEGylation Reagents
Other Product Names / Synonyms: 2-(2-Methoxyethoxy)ethanol, Methyl Carbitol, mPEG2-Alcohol
Formula:
C₅H₁₂O₃ MW: 120.15 g/mol Boiling Pt: 193 Melting Pt: -65 Density: 1.02 (20 °C) Flash Pt: 105 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002871 CAS Number: 111-77-3 EINECS: 203-906-6 Merck Index: 12,06116 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...