3-Methoxypyrocatechol ≥98.0%

Supplier: TCI America
Warning

Synonyms: 2,3-Dihydroxyanisole, 3-Methoxycatechol, 3-Methoxybenzene-1,2-diol

M0524-25G M0524-500G
TCM0524-025GEA 137.46 CAD
TCM0524-025G TCM0524-500G
3-Methoxypyrocatechol ≥98.0%
3-Methoxypyrocatechol

CAS Number: 934-00-9
MDL Number: MFCD00002191
Molecular Formula: C7H8O3
Molecular Weight: 140.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 164
Melting point (°C): 42
Flash Point (°C): 110
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 1,2-Dihydroxy-3-methoxybenzene, Pyrogallol 1-Monomethyl Ether

Formula: C₇H₈O₃
Boiling Pt: 164 °C
Melting Pt: 42 °C
Density: 1.2 g/cm³ (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00002191
CAS Number: 934-00-9
EINECS: 213-276-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=205ab466c30c83dfe7f38b979b3916ba77ce0f5f&vid=205ab466c30c83dfe7f38b979b3916ba77ce0f5f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR