2-Methylbenzothiazole ≥98.0%

Supplier: TCI America

M0516-25G M0516-500G M0516-100G
TCM0516-025GEA 194.34 CAD
TCM0516-025G TCM0516-500G TCM0516-100G
2-Methylbenzothiazole ≥98.0%
2-Methylbenzothiazole

CAS Number: 120-75-2
MDL Number: MFCD00005794
Molecular Formula: C8H7NS
Molecular Weight: 149.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 238
Flash Point (°C): 123
Freezing point (°C): 14
Specific Gravity (20/20): 1.18

Formula: C₈H₇NS
MW: 149.22 g/mol
Boiling Pt: 238 °C
Melting Pt: 12 °C
Density: 1.18 (20 °C)
Flash Pt: 123 °C
MDL Number: MFCD00005794
CAS Number: 120-75-2
EINECS: 204-423-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5262cc39bd3d850e0be5d08feb7c57958921a15e&vid=5262cc39bd3d850e0be5d08feb7c57958921a15e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR