(-)-Menthone ≥90.0%
Supplier: TCI America
M0513-25ML
M0513-500ML
M0513-100ML
TCM0513-025MLEA
64.27
CAD
TCM0513-025ML
TCM0513-500ML
TCM0513-100ML
(-)-Menthone ≥90.0%
(-)-Menthone
CAS Number: 14073-97-3
MDL Number: MFCD00001634
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 72
Specific Gravity (20/20): 0.89
Specific rotation [a]20/D: -19.5 deg (neat)
Product Categories: Bioscience, Monocyclic Monoterpenes
Other Product Names / Synonyms: (1R,4S)-(-)-p-Menthan-3-one
Formula:
C₁₀H₁₈O Boiling Pt: 88 °C Density: 0.89 (20 °C) Flash Pt: 72 °C |
MDL Number:
MFCD00001634 CAS Number: 14073-97-3 EINECS: 237-926-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...