2-Hydroxypropyl methacrylate (mixture of isomers) ≥97.0% stabilized

Supplier: TCI America

Synonyms: Propylene Glycol Monomethacrylate, HPMA, Hydroxypropyl methacrylate, Methacrylic acid 2-hydroxypropyl ester, Methacrylic acid hydroxypropyl ester

M0512-25ML M0512-500ML
TCM0512-025MLEA 31.6 CAD
TCM0512-025ML TCM0512-500ML
2-Hydroxypropyl methacrylate (mixture of isomers) ≥97.0% stabilized
2-Hydroxypropyl methacrylate (mixture of isomers, methacrylic acid, monoester with propane-1,2-diol)

(stabilized with MEHQ)


Form: Clear Liquid
Flash Point (°C): 104
Specific Gravity (20/20): 1.03

Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Density: 1.03 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00004536
CAS Number: 27813-02-1
EINECS: 248-666-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d17abbe07af252923a9f2017daf2cd7c28c9c5d&vid=1d17abbe07af252923a9f2017daf2cd7c28c9c5d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR