Metanil yellow

Supplier: TCI America

M0490-25G M0490-500G
TCM0490-025GEA 57.8 CAD
TCM0490-025G TCM0490-500G
Metanil yellow
Metanil Yellow

CAS Number: 587-98-4
MDL Number: MFCD00007486
Molecular Formula: C18H15N3O3S
Molecular Weight: 375.38
Form: Crystal
Color: Pale Yellow


Other Product Names / Synonyms: Metanil Yellow

Formula: C₁₈H₁₄N₃NaO₃S
MDL Number: MFCD00007486
CAS Number: 587-98-4
EINECS: 209-608-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc299578c6a67921e4ffc554f9935f799c962f11&vid=cc299578c6a67921e4ffc554f9935f799c962f11&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR