Ethyl myristate ≥98.0%

Supplier: TCI America

Synonyms: Tetradecanoic acid ethyl ester, Tetradecylic acid ethyl ester, Ethyl tetradecanoate, Myristic acid ethyl ester

M0479-25ML M0479-500ML M0479-100ML
TCM0479-025MLEA 36.34 CAD
TCM0479-025ML TCM0479-500ML TCM0479-100ML
Ethyl myristate ≥98.0%
Ethyl myristate

CAS Number: 124-06-1
MDL Number: MFCD00008984
Molecular Formula: C16H32O2
Molecular Weight: 256.43
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 309
Flash Point (°C): 167
Freezing point (°C): 12
Specific Gravity (20/20): 0.86


Product Categories: Bioscience, Lipids
Other Product Names / Synonyms: Myristic Acid Ethyl Ester

Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 309 °C
Density: 0.86 (20 °C)
Flash Pt: 167 °C
MDL Number: MFCD00008984
CAS Number: 124-06-1
EINECS: 204-675-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8aaee6c6f8ab93955479e7d733cb3864dfacd6b&vid=f8aaee6c6f8ab93955479e7d733cb3864dfacd6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR