Myristic acid ≥99.0% (by titrimetric analysis)

Supplier: TCI America
Warning

M0476-25G M0476-500G
TCM0476-25GEA 42.63 CAD
TCM0476-25G TCM0476-500G
Myristic acid ≥99.0% (by titrimetric analysis)
Myristic acid

CAS Number: 544-63-8
MDL Number: MFCD00002744
Molecular Formula: C14H28O2
Molecular Weight: 228.38
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Boiling point (°C): 326
Melting point (°C): 54
Flash Point (°C): 155


Product Categories: Materials Chemistry, Derivatizing Reagents for Solubility Improvement, Pressure & Heat Sensitive Recording Materials, Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: Tetradecanoic Acid

Formula: C₁₄H₂₈O₂
Boiling Pt: 326 °C
Melting Pt: 54 °C
Density: 0.8622 g/cm³ (20 °C)
Flash Pt: 155 °C
Storage Temperature: Freezer
MDL Number: MFCD00002744
CAS Number: 544-63-8
EINECS: 208-875-2
Merck Index: 12,06416
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cee7a7ce7abc7eee84c9034eac608e095fc8f3cb&vid=90c0929a17b60cf7a8c437035069905a786d1abe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR