Murexide

Supplier: TCI America

M0474-1G M0474-5G M0474-25G
TCM0474-001GEA 39.5 CAD
TCM0474-001G TCM0474-5G TCM0474-025G
Murexide
Murexide

CAS Number: 3051-09-0
MDL Number: MFCD00012777
Molecular Formula: C8H8N6O6
Molecular Weight: 284.19
Form: Crystal
Color: Deep Red


Other Product Names / Synonyms: Ammonium Purpurate

Formula: C₈H₈N₆O₆
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012777
CAS Number: 3051-09-0
EINECS: 221-266-6
Merck Index: 13,06330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b93691b6275560c0c5bf6d44836c4a8871128ea6&vid=b93691b6275560c0c5bf6d44836c4a8871128ea6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR