Mucic acid ≥97.0% (by titrimetric analysis)
Supplier: TCI America
M0466-500G
M0466-25G
TCM0466-500GEA
257.54
CAD
TCM0466-500G
TCM0466-025G
Mucic acid ≥97.0% (by titrimetric analysis)
Mucic acid
CAS Number: 526-99-8
MDL Number: MFCD00004239
Molecular Formula: C6H10O8
Molecular Weight: 210.14
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 210
Flash Point (°C): 100
Other Product Names / Synonyms: 2,3,4,5-Tetrahydroxyadipic Acid, Galactaric Acid
Formula:
C₆H₁₀O₈ Melting Pt: 210 °C Density: 1.79 g/cm³ (20 °C) Flash Pt: 100 °C |
MDL Number:
MFCD00004239 CAS Number: 526-99-8 EINECS: 208-404-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...