DL-Methionine 99,0

Supplier: TCI America

Synonyms: 2-Amino-4-(methylthio)butanoic acid, (±)-2-Amino-4-(methylthio)butyric acid, Methioninum racemicum, Methionine racematic, H-DL-Met-OH, (±)-Methionine, (±)-2-Amino-4-(methylthio)butanoic acid

M0463-500G M0463-25G
TCM0463-500GEA 213.16 CAD
TCM0463-500G TCM0463-025G
DL-Methionine 99,0
DL-Methionine

CAS Number: 59-51-8
MDL Number: MFCD00063096
Molecular Formula: C5H11NO2S
Molecular Weight: 149.21
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal


Product Categories: Bioscience, Proteinogenic Amino Acids
Other Product Names / Synonyms: H-DL-Met-OH

Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
Melting Pt: 136
Density: 1.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063096
CAS Number: 59-51-8
EINECS: 200-432-1
Merck Index: 13,06004
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b86b33aba6043ba7db47a7e44626516a56edbc7a&vid=b86b33aba6043ba7db47a7e44626516a56edbc7a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR