Methylurea ≥98.0% (by total nitrogen basis)

Supplier: TCI America

M0455-500G M0455-25G
TCM0455-500GEA 132.64 CAD
TCM0455-500G TCM0455-025G
Methylurea ≥98.0% (by total nitrogen basis)
Methylurea

CAS Number: 598-50-5
MDL Number: MFCD00007950
Molecular Formula: C2H6N2O
Molecular Weight: 74.08
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 99

Formula: C₂H₆N₂O
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 99 °C
Density: 1.204 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007950
CAS Number: 598-50-5
EINECS: 209-935-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=537efc41026055837f298d7da17270caf9861419&vid=537efc41026055837f298d7da17270caf9861419&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR