3,5-Dihydroxytoluene monohydrate ≥98.0%

Supplier: TCI America
Warning

Synonyms: 5-Methylresorcinol monohydrate, Orcinol monohydrate, 5-Methylbenzene-1,3-diol hydrate

M0426-5G M0426-25G
TCM0426-5GEA 82.16 CAD
TCM0426-5G TCM0426-025G
3,5-Dihydroxytoluene monohydrate ≥98.0%
3,5-Dihydroxytoluene monohydrate

CAS Number: 6153-39-5
MDL Number: MFCD00149092
Molecular Formula: C7H8O2
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 59


Other Product Names / Synonyms: 1,3-Dihydroxy-5-methylbenzene, 3,5-Dihydroxytoluene, Orcinol

Formula: C₇H₈O₂·1H₂O
MW: 142.15 g/mol
Boiling Pt: 290
Melting Pt: 59 °C
Density: 1.29 g/cm³ (4 °C)
Flash Pt: 159
Storage Temperature: Ambient
MDL Number: MFCD00149092
CAS Number: 6153-39-5
EINECS: 207-984-2
Merck Index: 12,06995
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=760cc40d97604d9134fc7d21fd68a91d05570b63&vid=760cc40d97604d9134fc7d21fd68a91d05570b63&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR