Methyl red

Supplier: TCI America

M0421-25G M0421-1G
TCM0421-025GEA 238.85 CAD
TCM0421-025G TCM0421-001G
Methyl red
Methyl red

CAS Number: 493-52-7
MDL Number: MFCD00002425
Molecular Formula: C15H15N3O2
Molecular Weight: 269.30
Form: Crystal
Color: Deep Red
Melting point (°C): 182
Lambda max.: 410 nm


Other Product Names / Synonyms: 4-Dimethylaminoazobenzene-2'-carboxylic Acid, Acid Red 2

Formula: C₁₅H₁₅N₃O₂
Melting Pt: 182 °C
Density: 1.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002425
CAS Number: 493-52-7
EINECS: 207-776-1
Merck Index: 12,06199
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6f65b0d2c9aab4635f969772d5a834b022c0492&vid=b6f65b0d2c9aab4635f969772d5a834b022c0492&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR