8-Methylquinoline ≥99.0%

Supplier: TCI America
Warning

M0419-25G M0419-5G
TCM0419-025GEA 132.72 CAD
TCM0419-025G TCM0419-5G
8-Methylquinoline ≥99.0%
8-Methylquinoline

CAS Number: 611-32-5
MDL Number: MFCD00006810
Molecular Formula: C10H9N
Molecular Weight: 143.19
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 248
Melting point (°C): -80
Flash Point (°C): 105
Specific Gravity (20/20): 1.08

Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 248 °C
Melting Pt: –80 °C
Density: 1.08 (20 °C)
Flash Pt: 105 °C
Storage Temperature: Ambient
MDL Number: MFCD00006810
CAS Number: 611-32-5
EINECS: 210-264-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7012bfdd23c84c699ac5505df1c0634201b5019b&vid=5802672698b566d80553e37cd08791d34fffe406&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR