N-Methyl-2-pyrrolidone (NMP) ≥99.0%

Supplier: TCI America
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone

M0418-500ML M0418-25ML M0418-100ML
TCM0418-500MLEA 85.3 CAD
TCM0418-500ML TCM0418-025ML TCM0418-100ML
N-Methyl-2-pyrrolidone (NMP) ≥99.0%
N-Methyl-2-pyrrolidone (NMP)

CAS Number: 872-50-4
MDL Number: MFCD00003193
Molecular Formula: C5H9NO
Molecular Weight: 99.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 202
Melting point (°C): -24
Flash Point (°C): 95
Specific Gravity (20/20): 1.03
Lambda max.: 283 nm (MeOH)


Product Categories: Materials Chemistry, Polyimide Raw Materials
Other Product Names / Synonyms: 1-Methyl-2-pyrrolidinone, NMP

Formula: C₅H₉NO
Boiling Pt: 202 °C
Melting Pt: –24 °C
Density: 1.03 (20 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=150f8dd99c84b55e9507e4974447c260f6c402d1&vid=150f8dd99c84b55e9507e4974447c260f6c402d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR