6-Methylquinoline ≥98.0% (by GC, titration analysis)
Supplier: TCI America
M0416-25G
M0416-5G
TCM0416-025GEA
110.6
CAD
TCM0416-025G
TCM0416-5G
6-Methylquinoline ≥98.0% (by GC, titration analysis)
6-Methylquinoline
CAS Number: 91-62-3
MDL Number: MFCD00006804
Molecular Formula: C10H9N
Molecular Weight: 143.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 259
Flash Point (°C): 110
Specific Gravity (20/20): 1.07
Formula:
C₁₀H₉N MW: 143.19 g/mol Boiling Pt: 259 °C Melting Pt: –22.2 °C Density: 1.07 (20 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006804 CAS Number: 91-62-3 EINECS: 202-084-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...