1-Methyl-2-pyridone ≥99.0%
Supplier: TCI America
|
Warning
|
Synonyms:
1-Methylpyridin-2-ol, N-Methylpyridin-2-ol, 1-Methylpyridin-2(1H)-one, N-Methyl-2-pyridone
M0412-25G
M0412-10G
TCM0412-25GEA
355.5
CAD
TCM0412-25G
TCM0412-010G
1-Methyl-2-pyridone ≥99.0%
1-Methyl-2-pyridone
CAS Number: 694-85-9
MDL Number: MFCD00006520
Molecular Formula: C6H7NO
Molecular Weight: 109.13
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 252
Flash Point (°C): 113
Specific Gravity (20/20): 1.13
Formula:
C₆H₇NO MW: 109.13 g/mol Boiling Pt: 250 Melting Pt: 33 Density: 1.13 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00006520 CAS Number: 694-85-9 EINECS: 211-777-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...