(Methylsulfonyl)benzene ≥97.0%

Supplier: TCI America

M0401-25G M0401-500G
TCM0401-025GEA 151.68 CAD
TCM0401-025G TCM0401-500G
(Methylsulfonyl)benzene ≥97.0%
(Methylsulphonyl)benzene

CAS Number: 3112-85-4
MDL Number: MFCD00014741
Molecular Formula: C7H8O2S
Molecular Weight: 156.20
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 88

Formula: C₇H₈O₂S
Melting Pt: 88 °C
MDL Number: MFCD00014741
CAS Number: 3112-85-4
EINECS: 221-476-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c268e10ab2d0b528d3d7722cbd504ef33f5e8fa&vid=0c268e10ab2d0b528d3d7722cbd504ef33f5e8fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR