(±)-2-Methyl-2,4-pentanediol ≥99.0%
Supplier: TCI America
|
Warning
|
M0384-500ML
M0384-25ML
TCM0384-500MLEA
63.17
CAD
TCM0384-500ML
TCM0384-025ML
(±)-2-Methyl-2,4-pentanediol ≥99.0%
(±)-2-Methyl-2,4-pentanediol
CAS Number: 107-41-5
MDL Number: MFCD00004547
Molecular Formula: C6H14O2
Molecular Weight: 118.18
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 197
Flash Point (°C): 98
Specific Gravity (20/20): 0.92
Other Product Names / Synonyms: 2,4-Dihydroxy-2-methylpentane, Hexylene Glycol
Formula:
C₆H₁₄O₂ Boiling Pt: 197 °C Melting Pt: –40 °C Density: 0.92 (20 °C) Flash Pt: 98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004547 CAS Number: 107-41-5 EINECS: 203-489-0 Merck Index: 13,04730 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...