Diethyl methylmalonate ≥98.0%

Supplier: TCI America

Synonyms: Diethyl 2-methylmalonate

M0366-500ML M0366-25ML M0366-100ML
TCM0366-500MLEA 200.66 CAD
TCM0366-500ML TCM0366-025ML TCM0366-100ML
Diethyl methylmalonate ≥98.0%
Diethyl methylmalonate

CAS Number: 609-08-5
MDL Number: MFCD00009162
Molecular Formula: C8H14O4
Molecular Weight: 174.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 199
Flash Point (°C): 82
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: Methylmalonic Acid Diethyl Ester

Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 199 °C
Density: 1.02 (20 °C)
Flash Pt: 82 °C
MDL Number: MFCD00009162
CAS Number: 609-08-5
EINECS: 210-175-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=060c0e09dbeea5cb73d85b9621b289eac1362512&vid=060c0e09dbeea5cb73d85b9621b289eac1362512&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR