2-Hydroxyisobutyric acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M0360-25G M0360-500G M0360-100G
TCM0360-025GEA 41.08 CAD
TCM0360-025G TCM0360-500G TCM0360-100G
2-Hydroxyisobutyric acid ≥98.0% (by titrimetric analysis)
2-Hydroxyisobutyric acid

CAS Number: 594-61-6
MDL Number: MFCD00004459
Molecular Formula: C4H8O3
Molecular Weight: 104.11
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 212
Melting point (°C): 80


Other Product Names / Synonyms: 2-Methyllactic Acid

Formula: C₄H₈O₃
Boiling Pt: 212 °C
Melting Pt: 80 °C
MDL Number: MFCD00004459
CAS Number: 594-61-6
EINECS: 209-848-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f110a4cd792bf2c170873ee2f57959a51eae4744&vid=f110a4cd792bf2c170873ee2f57959a51eae4744&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR