2-(Methylthio)ethanol ≥98.0%

Supplier: TCI America

M0358-25ML M0358-100ML
TCM0358-025MLEA 123.71 CAD
TCM0358-025ML TCM0358-100ML
2-(Methylthio)ethanol ≥98.0%
2-(Methylthio)ethanol

CAS Number: 5271-38-5
MDL Number: MFCD00002908
Molecular Formula: C3H8OS
Molecular Weight: 92.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 81
Flash Point (°C): 70
Specific Gravity (20/20): 1.06


Other Product Names / Synonyms: 2-Hydroxyethyl Methyl Sulfide

Formula: C₃H₈OS
Boiling Pt: 81 °C
Density: 1.06 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00002908
CAS Number: 5271-38-5
EINECS: 226-090-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca3e79e7a05e973ad065bec6d2e9598b6baa42a0&vid=ca3e79e7a05e973ad065bec6d2e9598b6baa42a0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR